Skip to Content
Merck
Nomegestrol acetate European Pharmacopoeia (EP) Reference Standard

Nomegestrol acetate

Nomegestrol acetate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
17-Acetoxy-6-methyl-19-norpregna-4,6-diene-3,20-dione, NOMAc, Org 10486-0
Empirical Formula (Hill Notation):
C23H30O4
CAS Number:
Molecular Weight:
370.48

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing