![N,N′-(o-Phenylene)dimaleimide 99%](/deepweb/assets/sigmaaldrich/product/structures/182/617/e5ac9bb7-87d9-49a3-8b46-e246ded62e09/640/e5ac9bb7-87d9-49a3-8b46-e246ded62e09.png)
N,N′-(o-Phenylene)dimaleimide
![N,N′-(o-Phenylene)dimaleimide 99%](/deepweb/assets/sigmaaldrich/product/structures/182/617/e5ac9bb7-87d9-49a3-8b46-e246ded62e09/640/e5ac9bb7-87d9-49a3-8b46-e246ded62e09.png)
Synonym(s):
1,2-Dimaleimidobenzene
Empirical Formula (Hill Notation):
C14H8N2O4
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing