Skip to Content
Merck
N,N′,N′′-Tritosyldiethylenetriamine ≥99.0%

N,N′,N′′-Tritosyldiethylenetriamine

N,N′,N′′-Tritosyldiethylenetriamine ≥99.0%
Linear Formula:
(CH3C6H4SO2NHCH2CH2)2NSO2C6H4CH3
CAS Number:
Molecular Weight:
565.73
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing