Skip to Content
Merck
N,N′-Hexamethylene bis(acetamide) 98%

N,N′-Hexamethylene bis(acetamide)

N,N′-Hexamethylene bis(acetamide) 98%
Synonym(s):
N,N′-Diacetyl-1,6-hexanediamine, HMBA
Linear Formula:
CH3CONH(CH2)6NHCOCH3
CAS Number:
Molecular Weight:
200.28

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing