
N,N-Dimethylformamide dibutyl acetal

Synonym(s):
1,1-Dibutoxy-N,N-dimethylmethylamine, 1,1-Dibutoxytrimethylamine
Linear Formula:
(CH3)2NCH(OCH2CH2CH2CH3)2
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing