Skip to Content
Merck
N,N-Dimethyl-2-chloroacetoacetamide Wacker Chemie AG, ≥95% (GC)

N,N-Dimethyl-2-chloroacetoacetamide

N,N-Dimethyl-2-chloroacetoacetamide Wacker Chemie AG, ≥95% (GC)
Synonym(s):
2-Chloro-N,N-dimethylacetoacetamide
Linear Formula:
CH3COCHClCON(CH3)2
CAS Number:
Molecular Weight:
163.60

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing