Skip to Content
Merck
N-Methyl-N-(2-pyridyl)formamide 99%

N-Methyl-N-(2-pyridyl)formamide

N-Methyl-N-(2-pyridyl)formamide 99%
Synonym(s):
N-Formyl-N-methylaminopyridine, Meyers’ Reagent
Empirical Formula (Hill Notation):
C7H8N2O
CAS Number:
Molecular Weight:
136.15
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing