
N-Methyl-1-(methylthio)-2-nitroethenamine

Linear Formula:
O2NCH=C(SCH3)NHCH3
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing