Skip to Content
Merck
N-Boc-D-glucosamine 97%

N-Boc-D-glucosamine

N-Boc-D-glucosamine 97%
Synonym(s):
N-(tert-Butoxycarbonyl)-D-glucosamine
Empirical Formula (Hill Notation):
C11H21NO7
CAS Number:
Molecular Weight:
279.29

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing