(N-Boc-β-amino)-Ala-OH
Synonym(s):
Boc-Dap-OH, (S)-2-[(tert-Butoxycarbonyl)amino]-3-aminopropionic acid, (S)-3-Amino-2-(tert-butoxycarbonyl)aminopropionic acid, (S)-3-Amino-2-(tert-butoxycarbonylamino)propanoic acid, 3-Amino-(tert-butoxycarbonyl)-L-alanine, Nα-BOC-(S)-β-aminoalanine, Nα-Boc-L-β-aminoalanine, N2-(tert-Butoxycarbonyl)-(S)-2,3-diaminopropionic acid, N2-tert-Butoxycarbonyl-L-2,3-diaminopropionic acid, Nα-Boc-L-2,3-diaminopropionic acid, Boc-Dpr-OH
Empirical Formula (Hill Notation):
C8H16N2O4
CAS Number:
Molecular Weight:
204.22
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing