Skip to Content
Merck
N-Boc-2-piperidinecarboxylic acid 98%

N-Boc-2-piperidinecarboxylic acid

N-Boc-2-piperidinecarboxylic acid 98%
Synonym(s):
(±)-1-Boc-piperidine-2-carboxylic acid, N-Boc-DL-pipecolinic acid, Boc-DL-Pip-OH
Empirical Formula (Hill Notation):
C11H19NO4
CAS Number:
Molecular Weight:
229.27

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing