Skip to Content
Merck
N-Boc-1H-pyrazole-1-carboxamidine 97%

N-Boc-1H-pyrazole-1-carboxamidine

N-Boc-1H-pyrazole-1-carboxamidine 97%
Empirical Formula (Hill Notation):
C9H14N4O2
CAS Number:
Molecular Weight:
210.23

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing