
Nalidixic acid impurity C

Synonym(s):
Diethyl ethoxymethylenemalonate, DEEMM, Ethoxymethylenemalonic acid diethyl ester
Linear Formula:
C2H5OCH=C(COOC2H5)2
CAS Number:
Molecular Weight:
216.23
Products
Boiling Point (°C)
Formula Weight
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing