N6-Phenoxyacetyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine
Empirical Formula (Hill Notation):
C39H37N5O7
CAS Number:
Molecular Weight:
687.74
Products
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing