Skip to Content
Merck
Morphine 6-β-D-glucuronide hydrate ≥90% (HPLC)

Morphine 6-β-D-glucuronide hydrate

Morphine 6-β-D-glucuronide hydrate ≥90% (HPLC)
Synonym(s):
M6G
Empirical Formula (Hill Notation):
C23H27NO9 · 2H2O
CAS Number:
Molecular Weight:
497.49

Products

Product Category
Brand
Formula Weight
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing