
Monoclonal Anti-Breast Cancer Resistance Protein antibody produced in mouse

Synonym(s):
Anti-ABCG2, Anti-BCRP
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing