Skip to Content
Merck
(−)-mono-(1R)-Menthyl phthalate 99%

(−)-mono-(1R)-Menthyl phthalate

(−)-mono-(1R)-Menthyl phthalate 99%
Empirical Formula (Hill Notation):
C18H24O4
CAS Number:
Molecular Weight:
304.38

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing