
Molybdenum sulfide concentrate

Synonym(s):
Molybdenum(IV) sulfide, Molybdenum disulfide
Empirical Formula (Hill Notation):
MoS2
CAS Number:
Molecular Weight:
160.07
Products
Formula Weight
Manufacturer
Physical Form
Quality Segment
Application
Showing 1-1 of 1