Skip to Content
Merck
Micro particles based on melamine resin, carboxylate-modified, rhodamine B-marked size: 10 μm

Micro particles based on melamine resin, carboxylate-modified, rhodamine B-marked

Micro particles based on melamine resin, carboxylate-modified, rhodamine B-marked size: 10 μm
Synonym(s):
Melamine-formaldehyde resin

Products

Brand
Physical Form
Format
Application
Particle Size (µm)
Reference Material Type
Special Grade
Showing 1-1 of 1