Skip to Content
Merck
Methyl pentacosanoate analytical standard

Methyl pentacosanoate

Methyl pentacosanoate analytical standard
Linear Formula:
CH3(CH2)23COOCH3
CAS Number:
Molecular Weight:
396.69
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing