Skip to Content
Merck
Methyl L-3-phenyllactate ≥97.0% (HPLC)

Methyl L-3-phenyllactate

Methyl L-3-phenyllactate ≥97.0% (HPLC)
Synonym(s):
Methyl (S)-2-hydroxy-3-phenylpropionate
Empirical Formula (Hill Notation):
C10H12O3
CAS Number:
Molecular Weight:
180.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing