Skip to Content
Merck
Methyl 6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside ≥97.0% (TLC)

Methyl 6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside

Methyl 6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside ≥97.0% (TLC)
Synonym(s):
6-O-(N-Heptylcarbamoyl)-methyl-α-D-glucopyranoside, HECAMEG
Empirical Formula (Hill Notation):
C15H29NO7
CAS Number:
Molecular Weight:
335.39

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing