Skip to Content
Merck
Methyl 4-oxo-3-piperidinecarboxylate hydrochloride 97%

Methyl 4-oxo-3-piperidinecarboxylate hydrochloride

Methyl 4-oxo-3-piperidinecarboxylate hydrochloride 97%
Synonym(s):
3-Methoxycarbonyl-4-piperidone hydrochloride
Empirical Formula (Hill Notation):
C7H11NO3 · HCl
CAS Number:
Molecular Weight:
193.63
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing