Methyl 4-hydroxybenzoate-ring-13C6 solution
Synonym(s):
4-Hydroxybenzoic acid methyl ester-ring-13C6 solution, Methyl paraben-ring-13C6 solution
Empirical Formula (Hill Notation):
13C6C2H8O3
Molecular Weight:
158.10
EC Number:
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing