![Methyl 4-fluorobenzoylacetate 95%](/deepweb/assets/sigmaaldrich/product/structures/172/334/6fcbe487-5be0-4e96-a547-29d41b829825/640/6fcbe487-5be0-4e96-a547-29d41b829825.png)
Methyl 4-fluorobenzoylacetate
![Methyl 4-fluorobenzoylacetate 95%](/deepweb/assets/sigmaaldrich/product/structures/172/334/6fcbe487-5be0-4e96-a547-29d41b829825/640/6fcbe487-5be0-4e96-a547-29d41b829825.png)
Synonym(s):
Methyl 3-(4-fluorophenyl)-3-oxopropionate
Linear Formula:
FC6H4COCH2CO2CH3
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing