Skip to Content
Merck
Methyl 3-(trimethylsilyl)-4-pentenoate 97%

Methyl 3-(trimethylsilyl)-4-pentenoate

Methyl 3-(trimethylsilyl)-4-pentenoate 97%
Linear Formula:
H2C=CHCH[Si(CH3)3]CH2CO2CH3
CAS Number:
Molecular Weight:
186.32

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing