Skip to Content
Merck
Methyl 3-bromo-4-methoxybenzoate 98%

Methyl 3-bromo-4-methoxybenzoate

Methyl 3-bromo-4-methoxybenzoate 98%
Synonym(s):
3-Bromo-4-methoxybenzoic acid methyl ester
Linear Formula:
CH3OC6H3(Br)CO2CH3
CAS Number:
Molecular Weight:
245.07

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing