Skip to Content
Merck
Methyl 2,6-dichloropyridine-4-carboxylate 97%

Methyl 2,6-dichloropyridine-4-carboxylate

Methyl 2,6-dichloropyridine-4-carboxylate 97%
Synonym(s):
Methyl 2,6-dichloroisonicotinate
Empirical Formula (Hill Notation):
C7H5Cl2NO2
CAS Number:
Molecular Weight:
206.03
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing