Skip to Content
Merck
Methyl 1-cyclopentene-1-carboxylate 97%

Methyl 1-cyclopentene-1-carboxylate

Methyl 1-cyclopentene-1-carboxylate 97%
Linear Formula:
C5H7CO2CH3
CAS Number:
Molecular Weight:
126.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing