Skip to Content
VN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methanol-d
4
Linear Formula:
CD
3
OD
Molecular Weight:
36.07
EC Number:
212-378-6
Products
Brand
Supelco (2)
Formula Weight
Physical Form
Format
Isotope
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
721921
Methanol-d
4
,
NMR reference standard, 99.8 atom % D, NMR tube size 3 mm × 8 in.
Expand
View Pricing
721948
Methanol-d
4
,
NMR reference standard, 99.8 atom % D, NMR tube size 5 mm × 8 in.
Expand
View Pricing