Skip to Content
Merck
Mercury(I) chloride anhydrous, powder, 99.999% trace metals basis

Mercury(I) chloride

Mercury(I) chloride anhydrous, powder, 99.999% trace metals basis
Synonym(s):
Calomel, Mercurous chloride
Linear Formula:
Hg2Cl2
CAS Number:
Molecular Weight:
472.09
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Special Grade
Showing 1-2 of 2