Skip to Content
Merck
Menthyl anthranilate Pharmaceutical Secondary Standard; Certified Reference Material

Menthyl anthranilate

Menthyl anthranilate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Menthyl o-aminobenzoate, Menthyl anthranilate
Empirical Formula (Hill Notation):
C17H25NO2
CAS Number:
Molecular Weight:
275.39
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2