Skip to Content
Merck
Mefenamic acid impurity A European Pharmacopoeia (EP) Reference Standard

Mefenamic acid impurity A

Mefenamic acid impurity A European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2,3-Dimethylaniline, 1-Amino-2,3-dimethylbenzene, 2,3-Xylidine, 3-Amino-o-xylene, CN-Cbl
Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18

Products

Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing