Skip to Content
Merck
Mazindol ≥98% (TLC), powder

Mazindol

Mazindol ≥98% (TLC), powder
Synonym(s):
5-(4-Chlorophenyl)-2,5-dihydro-3H-imidazo[2,1-a]isoindol-5-ol
Empirical Formula (Hill Notation):
C16H13ClN2O
CAS Number:
Molecular Weight:
284.74
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing