Skip to Content
Merck
Luperox® 224, 2,4-Pentanedione peroxide solution ~34 wt. % in 4-hydroxy-4-methyl-2-pentanone and N-methyl-2-pyrrolidone

Luperox® 224, 2,4-Pentanedione peroxide solution

Luperox® 224, 2,4-Pentanedione peroxide solution ~34 wt. % in 4-hydroxy-4-methyl-2-pentanone and N-methyl-2-pyrrolidone
Synonym(s):
2,4-Pentanedione peroxide solution
Empirical Formula (Hill Notation):
C10H14O6
CAS Number:
Molecular Weight:
230.21

Products

Brand
Boiling Point (°C)
Formula Weight
Markush Class
Markush Group
Reagent Type