Skip to Content
Merck
L-Aspartic acid dimethyl ester hydrochloride 97%

L-Aspartic acid dimethyl ester hydrochloride

L-Aspartic acid dimethyl ester hydrochloride 97%
Linear Formula:
CH3O2CCH2CH(NH2)CO2CH3 · HCl
CAS Number:
Molecular Weight:
197.62

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing