
Isotianil

Synonym(s):
3,4-Dichloro-2′-cyano-1,2-thiazole-5-carboxanilide, 3,4-Dichloro-N-(2-cyanophenyl)-5-isothiazolecarboxamide
Empirical Formula (Hill Notation):
C11H5Cl2N3OS
CAS Number:
Molecular Weight:
298.15
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing