Skip to Content
Merck
Isosafrol mixture of cis and trans

Isosafrol

Isosafrol mixture of cis and trans
Synonym(s):
1,2-Methylenedioxy-4-propenylbenzene
Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing