![Isomethiozin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/278/562/e4802925-ba3a-4dd7-bb88-b58759e7667c/640/e4802925-ba3a-4dd7-bb88-b58759e7667c.png)
Isomethiozin
![Isomethiozin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/278/562/e4802925-ba3a-4dd7-bb88-b58759e7667c/640/e4802925-ba3a-4dd7-bb88-b58759e7667c.png)
Empirical Formula (Hill Notation):
C12H20N4OS
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing