
Isocyanate, polymer-bound

Synonym(s):
Methyl isocyanate bound to polystyrene
Empirical Formula (Hill Notation):
C26H25NO
CAS Number:
Molecular Weight:
367.48
Products
Brand
Formula Weight
Chain Type
Reaction Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing