Skip to Content
Merck
Indo 1-AM ~95% (HPLC)

Indo 1-AM

Indo 1-AM ~95% (HPLC)
Synonym(s):
4-(6-Carboxy-2-indolyl)-4′-methyl-2,2′-(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester
Empirical Formula (Hill Notation):
C47H51N3O22
CAS Number:
Molecular Weight:
1009.91

Products

Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing