
IAEA-N-2 (nitrogen isotopes in ammonium sulfate)

Synonym(s):
Ammonium sulfate, Mascagnite
Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Isotope
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing