Skip to Content
Merck
Hydroquinidine 4-chlorobenzoate 98%

Hydroquinidine 4-chlorobenzoate

Hydroquinidine 4-chlorobenzoate 98%
Synonym(s):
O-(4-Chlorobenzoyl)hydroquinidine, Dihydroquinidine 4-chlorobenzoate
Empirical Formula (Hill Notation):
C27H29ClN2O3
CAS Number:
Molecular Weight:
464.98

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing