Skip to Content
Merck
Glipizide Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material

Glipizide Related Compound B

Glipizide Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
6-Methyl-N-[2-(4-sulphamoylphenyl)ethyl]pyrazine-2-carboxamide
Empirical Formula (Hill Notation):
C14H16N4O3S
CAS Number:
Molecular Weight:
320.37

Products

Brand
Formula Weight
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1