Skip to Content
Merck
Furmecyclox PESTANAL®, analytical standard

Furmecyclox

Furmecyclox PESTANAL®, analytical standard
Synonym(s):
N-Cyclohexyl-N-methoxy-2,5-dimethyl-3-furamide
Empirical Formula (Hill Notation):
C14H21NO3
CAS Number:
Molecular Weight:
251.32
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing