Skip to Content
Merck
Follicle Stimulating Hormone from porcine pituitary

Follicle Stimulating Hormone from porcine pituitary

Follicle Stimulating Hormone from porcine pituitary
Synonym(s):
FSH, Follitropin, pFSH
CAS Number:

Products

Product Category
Brand
Biological Source
Gene Alias
Physical Form
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing