Skip to Content
Merck
Fmoc-Arg-OH ≥96.0%

Fmoc-Arg-OH

Fmoc-Arg-OH ≥96.0%
Synonym(s):
Nα-Fmoc-L-arginine, Nα-(9-Fluorenylmethoxycarbonyl)-L-arginine
Empirical Formula (Hill Notation):
C21H24N4O4
CAS Number:
Molecular Weight:
396.44

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing