
Fluorene-2-boronic acid

Synonym(s):
9H-Fluoren-2-yl-boronic acid
Linear Formula:
C13H9B(OH)2
CAS Number:
Molecular Weight:
210.04
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing