Fleroxacin
Synonym(s):
6,8-Difluoro-1-(2-fluoroethyl)-1,4-dihydro-7-(4-methylpiperazino)-4-oxo-3-quinolinecarboxylic acid, 6,8-Difluoro-1-(2-fluoroethyl)1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, AM-833, Megalocin, Megalone
Empirical Formula (Hill Notation):
C17H18F3N3O3
CAS Number:
Molecular Weight:
369.34
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing