
Fenoxaprop

Synonym(s):
2-{4-[(6-Chlorobenzoxazol-2-yl)oxy]phenoxy}propionic acid
Empirical Formula (Hill Notation):
C16H12ClNO5
CAS Number:
Molecular Weight:
333.72
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing